Assay Details
Gene Symbol: Alox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012822.1 | NP_036954.1 | 7 - 8 | 1068 | 147 | |
XM_006237140.2 | XP_006237202.1 | 7 - 8 | 1105 | 147 | ||
GenBank mRNA | J03960.1 | - | 7 - 8 | 1068 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
