Assay Details
Gene Symbol: B4galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191976.1 | NP_001178905.1 | 15 - 16 | 2351 | 134 | |
XM_006237267.2 | XP_006237329.1 | 15 - 16 | 2654 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
