Assay Details
Gene Symbol: B4galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191976.1 | NP_001178905.1 | 16 - 17 | 2502 | 75 | |
XM_006237267.2 | XP_006237329.1 | 16 - 17 | 2805 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
