Assay Details
Gene Symbol: B4galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191976.1 | NP_001178905.1 | 18 - 19 | 2732 | 73 | |
XM_006237267.2 | XP_006237329.1 | 18 - 19 | 3035 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
