Assay Details
Gene Symbol: Pyroxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004234.1 | NP_001004234.1 | 8 - 9 | 915 | 71 | |
GenBank mRNA | BC079377.1 | - | 8 - 9 | 915 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
