Assay Details
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031633.3 | NP_113821.2 | 7 - 8 | 1143 | 61 | |
XM_006237417.2 | XP_006237479.1 | 5 - 6 | 1083 | 61 | ||
XM_006237421.2 | XP_006237483.1 | 6 - 7 | 1107 | 61 | ||
GenBank mRNA | U83112.1 | - | 7 - 8 | 1143 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
