Assay Details
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237416.2 | XP_006237478.1 | 3 - 4 | 902 | 83 | |
XM_006237417.2 | XP_006237479.1 | 3 - 4 | 902 | 83 | ||
XM_006237418.2 | XP_006237480.1 | 4 - 5 | 926 | 83 | ||
XM_006237419.2 | XP_006237481.1 | 3 - 4 | 902 | 83 | ||
XM_006237420.2 | XP_006237482.1 | 4 - 5 | 926 | 83 | ||
XM_006237421.2 | XP_006237483.1 | 4 - 5 | 926 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
