Assay Details
Gene Symbol: Slc25a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044280.2 | NP_001037745.2 | 8 - 9 | 1018 | 56 | |
GenBank mRNA | BC094517.1 | - | 8 - 9 | 963 | 7314749 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
