Assay Details
Gene Symbol: Ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106426.1 | NP_001099896.1 | 1 - 2 | 194 | 62 | |
GenBank mRNA | FQ224246.1 | - | 1 - 2 | 145 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
