Assay Details
Gene Symbol: Ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053702.3 | NP_446154.3 | 1 - 2 | 268 | 163 | |
XM_006232266.2 | XP_006232328.1 | 1 - 2 | 317 | 163 | ||
XM_008760903.1 | XP_008759125.1 | 1 - 2 | 319 | 163 | ||
GenBank mRNA | AF367448.1 | - | 1 - 2 | 167 | 163 | |
BC127462.1 | - | 1 - 2 | 273 | 163 | ||
DY317814.1 | - | 1 - 2 | 273 | 163 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC127462.1, XM_006232266.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
