Assay Details
Gene Symbol: Aggf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231820.2 | XP_006231882.1 | 2 - 3 | 614 | 136 | |
GenBank mRNA | BC099754.1 | - | 2 - 3 | 510 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
