Assay Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728994.4 | XP_002729040.2 | 5 - 6 | 1008 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
