Assay Details
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011946.1 | NP_001011946.1 | 11 - 12 | 1517 | 95 | |
GenBank mRNA | BC079376.1 | - | 11 - 12 | 1517 | 95 | |
FQ221834.1 | - | 6 - 7 | 641 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
