Assay Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760610.1 | XP_008758832.1 | 9 - 10 | 1533 | 113 | |
XM_008760611.1 | XP_008758833.1 | 8 - 9 | 1494 | 113 | ||
XM_008760612.1 | XP_008758834.1 | 10 - 11 | 4213 | 113 | ||
XM_008760613.1 | XP_008758835.1 | 9 - 10 | 4077 | 113 | ||
XM_008760614.1 | XP_008758836.1 | 9 - 10 | 1529 | 113 | ||
XM_008760615.1 | XP_008758837.1 | 8 - 9 | 1490 | 113 | ||
XM_008760616.1 | XP_008758838.1 | 5 - 6 | 3683 | 113 | ||
XM_008760617.1 | XP_008758839.1 | 5 - 6 | 564 | 113 | ||
GenBank mRNA | FQ232558.1 | - | 8 - 9 | 1113 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
