Assay Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231985.2 | XP_006232047.1 | 3 - 4 | 342 | 78 | |
XM_006231986.2 | XP_006232048.1 | 2 - 3 | 239 | 78 | ||
GenBank mRNA | FQ220090.1 | - | 3 - 4 | 291 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
