Assay Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007633.2 | NP_001007634.1 | 4 - 5 | 554 | 74 | |
NM_001162413.1 | NP_001155885.1 | 5 - 6 | 676 | 74 | ||
XM_006231985.2 | XP_006232047.1 | 5 - 6 | 622 | 74 | ||
XM_006231986.2 | XP_006232048.1 | 4 - 5 | 519 | 74 | ||
GenBank mRNA | BC079181.1 | - | 5 - 6 | 642 | 74 | |
FM123325.1 | - | 4 - 5 | 508 | 74 | ||
FQ220090.1 | - | 5 - 6 | 571 | 74 | ||
FQ220210.1 | - | 4 - 5 | 467 | 74 | ||
FQ230755.1 | - | 4 - 5 | 477 | 74 | ||
FQ235152.1 | - | 5 - 6 | 604 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
