Assay Details
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760700.1 | XP_008758922.1 | 3 - 4 | 287 | 89 | |
GenBank mRNA | X83579.1 | - | 3 - 4 | 133 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
