Assay Details
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760700.1 | XP_008758922.1 | 6 - 7 | 542 | 116 | |
GenBank mRNA | X83579.1 | - | 6 - 7 | 388 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
