Assay Details
Gene Symbol: Sgtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181629.3 | NP_853660.1 | 4 - 5 | 378 | 78 | |
XM_006231882.2 | XP_006231944.1 | 4 - 5 | 544 | 78 | ||
XM_006231883.2 | XP_006231945.1 | 4 - 5 | 519 | 78 | ||
GenBank mRNA | AF368280.1 | - | 4 - 5 | 379 | 78 | |
FQ211527.1 | - | 4 - 5 | 422 | 78 | ||
FQ212077.1 | - | 4 - 5 | 546 | 78 | ||
FQ212215.1 | - | 4 - 5 | 521 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
