Assay Details
Gene Symbol: Sgtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181629.3 | NP_853660.1 | 7 - 8 | 723 | 148 | |
XM_006231882.2 | XP_006231944.1 | 7 - 8 | 889 | 148 | ||
GenBank mRNA | AF368280.1 | - | 7 - 8 | 724 | 148 | |
FQ211527.1 | - | 7 - 8 | 767 | 148 | ||
FQ212077.1 | - | 7 - 8 | 891 | 148 | ||
FQ212215.1 | - | 7 - 8 | 866 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
