Assay Details
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC059113.1 | - | 4 - 5 | 616 | 112 | |
FN800625.1 | - | 4 - 5 | 614 | 112 | ||
FN800627.1 | - | 2 - 3 | 269 | 112 | ||
L11995.1 | - | 4 - 5 | 608 | 112 | ||
X60768.1 | - | 4 - 5 | 546 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
