Assay Details
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC059113.1 | - | 8 - 9 | 1266 | 108 | |
EV765751.1 | - | 4 - 5 | 671 | 108 | ||
FN800629.1 | - | 3 - 4 | 375 | 108 | ||
L11995.1 | - | 8 - 9 | 1258 | 108 | ||
X60768.1 | - | 8 - 9 | 1196 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
