Assay Details
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107650.2 | NP_001101120.2 | 9 - 10 | 900 | 74 | |
XM_008760726.1 | XP_008758948.1 | 9 - 10 | 962 | 74 | ||
XM_008760728.1 | XP_008758950.1 | 8 - 9 | 810 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
biological_process
response to UV
response to X-ray
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA repair
protein autoubiquitination
molecular_function
DNA helicase activity
ubiquitin-protein transferase activity
DNA-dependent ATPase activity
protein complex binding
protein polyubiquitination
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
biological_process
response to UV
response to X-ray
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA repair
protein autoubiquitination
molecular_function
DNA helicase activity
ubiquitin-protein transferase activity
DNA-dependent ATPase activity
protein complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P535959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137620 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P535959 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P535959 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P535959 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P535959 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18920 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P535953 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23416 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29459 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10812916 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812911 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812915 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812914 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812912 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812917 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812918 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812919 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812910 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812909 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10812906 | - | - |
Rat Genome 230 2.0 Array | 1379299_at | - | - |
Rat Genome 230 2.0 Array | 1379299_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379299_at | - | - |
Affymetrix HT Rat Focus Array | 1379299_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200001969.rn.1 | TC0200000343.rn.1 | RTA TC |
Rat Expression Set 230 | 1379299_PM_at | - | - |
Rat Expression Set 230 | 1379299_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10812906 | - | - |
Rat Genome U34 Set | rc_AA800156_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21709728 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366313 | - | - |
