Assay Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012853.1 | NP_036985.1 | 3 - 4 | 375 | 153 | |
XM_006254793.2 | XP_006254855.1 | 4 - 5 | 914 | 153 | ||
XM_008772089.1 | XP_008770311.1 | 2 - 3 | 2040 | 153 | ||
XM_008772090.1 | XP_008770312.1 | 2 - 3 | 2040 | 153 | ||
GenBank mRNA | AJ011370.1 | - | 4 - 5 | 455 | 153 | |
AM712909.1 | - | 3 - 4 | 355 | 153 | ||
AM712910.1 | - | 3 - 4 | 355 | 153 | ||
AM712911.1 | - | 3 - 4 | 355 | 153 | ||
U20906.1 | - | 3 - 4 | 375 | 153 | ||
U20907.1 | - | 3 - 4 | 375 | 153 | ||
Z48153.1 | - | 1 - 2 | 73 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
