Assay Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254793.2 | XP_006254855.1 | 1 - 2 | 510 | 135 | |
GenBank mRNA | AJ011370.1 | - | 1 - 2 | 51 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ011370.1, XM_006254793.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
