Assay Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ011370.1 | - | 6 - 7 | 1175 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
