Assay Details
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012853.1 | NP_036985.1 | 5 - 6 | 1091 | 76 | |
XM_008772089.1 | XP_008770311.1 | 4 - 5 | 2756 | 76 | ||
GenBank mRNA | AM712910.1 | - | 5 - 6 | 1071 | 76 | |
U20907.1 | - | 5 - 6 | 1091 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-activating G-protein coupled receptor signaling pathway
serotonin receptor signaling pathway
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
positive regulation of cell proliferation
regulation of appetite
cognition
G-protein coupled serotonin receptor activity
adenylate cyclase-activating G-protein coupled receptor signaling pathway
serotonin receptor signaling pathway
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
positive regulation of cell proliferation
regulation of appetite
cognition
G-protein coupled serotonin receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P108419 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P019097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41988 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019097 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108419 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019097 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108419 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P019097 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P108419 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P226802 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51543 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53483 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47205 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31790 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44949 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18718 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39243 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10802152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802153 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10802151 | - | - |
Rat Genome 230 2.0 Array | 1369533_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1391263_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391263_at | - | - |
Rat Genome 230 2.0 Array | 1369533_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369533_a_at | - | - |
Affymetrix HT Rat Focus Array | 1391263_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391263_at | - | - |
Affymetrix HT Rat Focus Array | 1369533_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800004793.rn.1 | TC1800000646.rn.1 | RTA TC |
Rat Expression Set 230 | 1369533_PM_a_at | - | - |
Rat Expression Set 230 | 1369533_a_at | - | - |
Rat Expression Set 230 | 1391263_at | - | - |
Rat Expression Set 230 | 1391263_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802151 | - | - |
Rat Genome U34 Set | U20907_at | - | - |
Rat Genome U34 Set | AJ011370_g_at | - | - |
Rat Genome U34 Set | AJ011370_at | - | - |
Rat Genome U34 Set | U20907_at | - | - |
Rat Genome U34 Set | AJ011370_at | - | - |
Rat Genome U34 Set | AJ011370_g_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21772855 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366668 | - | - |
RatRef-12 Expression BeadChip | ILMN_51684 | - | - |
