Assay Details
Gene Symbol: Ccbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772204.1 | XP_008770426.1 | 5 - 6 | 1497 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
