Assay Details
Gene Symbol: Sohlh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034961.1 | NP_001030133.1 | 1 - 2 | 79 | 84 | |
GenBank mRNA | BC104697.1 | - | 1 - 2 | 79 | 84 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC104697.1, NM_001034961.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
