Assay Details
Gene Symbol: Ccdc169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109184.1 | NP_001102654.1 | 4 - 5 | 363 | 127 | |
XM_006232368.2 | XP_006232430.1 | 4 - 5 | 1944 | 127 | ||
XM_008761003.1 | XP_008759225.1 | 4 - 5 | 1944 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
