Assay Details
Gene Symbol: Alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025407.1 | NP_001020578.1 | 2 - 3 | 244 | 70 | |
XM_006232333.1 | XP_006232395.1 | 2 - 3 | 479 | 70 | ||
GenBank mRNA | BC098039.1 | - | 2 - 3 | 244 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
