Assay Details
Gene Symbol: Mgst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106430.1 | NP_001099900.1 | 398 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
