Assay Details
Gene Symbol: Mgst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106430.1 | NP_001099900.1 | 482 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione biosynthetic process
response to organonitrogen compound
leukotriene biosynthetic process
response to lipopolysaccharide
positive regulation of catalytic activity
oxidation-reduction process
glutathione transferase activity
leukotriene-C4 synthase activity
glutathione peroxidase activity
enzyme activator activity
glutathione biosynthetic process
response to organonitrogen compound
leukotriene biosynthetic process
response to lipopolysaccharide
positive regulation of catalytic activity
oxidation-reduction process
glutathione transferase activity
leukotriene-C4 synthase activity
glutathione peroxidase activity
enzyme activator activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78761 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10641 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10641 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10641 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10641 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85786 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10815312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10815311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10815309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10815310 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10815308 | - | - |
Rat Genome 230 2.0 Array | 1372599_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372599_at | - | - |
Affymetrix HT Rat Focus Array | 1372599_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372599_at | - | - |
Rat Expression Set 230 | 1372599_at | - | - |
Rat Expression Set 230 | 1372599_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10815308 | - | - |
Rat Genome U34 Set | rc_AA955983_at | - | - |
Rat Genome U34 Set | rc_AA955983_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22341659 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370709 | - | - |
