Assay Details
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024869.1 | NP_001020040.1 | 3 - 4 | 730 | 64 | |
GenBank mRNA | BC097968.1 | - | 3 - 4 | 730 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
