Assay Details
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022252.2 | NP_071588.2 | 4 - 5 | 1442 | 127 | |
XM_008761058.1 | XP_008759280.1 | 4 - 5 | 1866 | 127 | ||
GenBank mRNA | BC078832.1 | - | 4 - 5 | 1442 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
