Assay Details
Gene Symbol: Elf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012181.2 | NP_001012181.1 | 3 - 4 | 1087 | 144 | |
NM_001033909.1 | NP_001029081.1 | 5 - 6 | 767 | 144 | ||
XM_006232325.2 | XP_006232387.1 | 6 - 7 | 969 | 144 | ||
XM_006232327.2 | XP_006232389.1 | 3 - 4 | 3155 | 144 | ||
XM_006232328.1 | XP_006232390.1 | 4 - 5 | 481 | 144 | ||
XM_006232329.1 | XP_006232391.1 | 4 - 5 | 445 | 144 | ||
XM_008760992.1 | XP_008759214.1 | 5 - 6 | 1352 | 144 | ||
GenBank mRNA | BC083585.1 | - | 6 - 7 | 919 | 7131145 | 144 |
BC083598.1 | - | 3 - 4 | 439 | 7132941 | 144 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
