Assay Details
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199091.2 | NP_954522.1 | 13 - 14 | 1550 | 74 | |
GenBank mRNA | BC063178.1 | - | 13 - 14 | 1554 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109445 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034876 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034876 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034876 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P525827 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P360325 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82403 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27727 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4945 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96241 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10816488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816495 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816492 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768174 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816497 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816494 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816493 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816489 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10768173 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10816487 | - | - |
Rat Genome 230 2.0 Array | 1371403_at | - | - |
Rat Genome 230 2.0 Array | 1371403_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371403_at | - | - |
Affymetrix HT Rat Focus Array | 1371403_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200009688.rn.1 | TC0200001540.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200009689.rn.1 | TC0200001540.rn.1 | RTA TC |
Rat Expression Set 230 | 1371403_PM_at | - | - |
Rat Expression Set 230 | 1371403_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10816487 | - | - |
Rat Genome U34 Set | rc_AA799545_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21614806 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351771 | - | - |
RatRef-12 Expression BeadChip | ILMN_52600 | - | - |
