Assay Details
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191847.1 | NP_001178776.1 | 15 - 16 | 1421 | 95 | |
GenBank mRNA | FQ211604.1 | - | 1438 | 95 | ||
FQ217493.1 | - | 1406 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
