Assay Details
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017020.3 | NP_058716.2 | 2 - 3 | 339 | 118 | |
XM_006232587.2 | XP_006232649.1 | 2 - 3 | 1773 | 118 | ||
XM_006232588.2 | XP_006232650.1 | 2 - 3 | 1772 | 118 | ||
XM_008761127.1 | XP_008759349.1 | 2 - 3 | 1768 | 118 | ||
GenBank mRNA | M58587.1 | - | 2 - 3 | 428 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
