Assay Details
Gene Symbol: Rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031092.1 | NP_112354.1 | 2 - 3 | 266 | 120 | |
GenBank mRNA | AF525280.1 | - | 2 - 3 | 266 | 120 | |
FQ222701.1 | - | 2 - 3 | 192 | 120 | ||
M83678.1 | - | 1 - 2 | 65 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
