Assay Details
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053550.1 | NP_446002.1 | 5 - 6 | 766 | 100 | |
XM_008768131.1 | XP_008766353.1 | 5 - 6 | 1045 | 100 | ||
GenBank mRNA | AF033355.1 | - | 5 - 6 | 766 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
