Assay Details
Gene Symbol: Slc13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031746.1 | NP_113934.1 | 4 - 5 | 639 | 80 | |
GenBank mRNA | AB001321.1 | - | 4 - 5 | 642 | 80 | |
AF058714.1 | - | 4 - 5 | 639 | 80 | ||
U51153.1 | - | 4 - 5 | 631 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
