Assay Details
Gene Symbol: Eif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033681.1 | NP_001028853.1 | 1 - 2 | 85 | 93 | |
XM_006246635.2 | XP_006246697.1 | 2 - 3 | 315 | 93 | ||
GenBank mRNA | BC101891.1 | - | 1 - 2 | 85 | 7930704 | 93 |
FQ213502.1 | - | 1 - 2 | 122 | 93 | ||
FQ219881.1 | - | 1 - 2 | 169 | 93 | ||
FQ220539.1 | - | 1 - 2 | 99 | 93 | ||
FQ226884.1 | - | 1 - 2 | 97 | 93 | ||
FQ229255.1 | - | 1 - 2 | 108 | 93 | ||
FQ231501.1 | - | 1 - 2 | 67 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC101891.1,NM_001033681.1,XM_006246635.2 transcripts.
This assay is in the 5'UTR region of the BC101891.1,NM_001033681.1,XM_006246635.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
