Assay Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012998.1 | NP_037130.1 | 10 - 11 | 1493 | 147 | |
GenBank mRNA | BC061857.1 | - | 10 - 11 | 1507 | 5623688 | 147 |
FQ209800.1 | - | 3 - 4 | 307 | 147 | ||
FQ220001.1 | - | 2 - 3 | 280 | 147 | ||
M21018.1 | - | 10 - 11 | 1371 | 147 | ||
X02918.1 | - | 10 - 11 | 1493 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
