Assay Details
Gene Symbol: P4hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012998.1 | NP_037130.1 | 3 - 4 | 528 | 67 | |
GenBank mRNA | BC061857.1 | - | 3 - 4 | 542 | 67 | |
M21018.1 | - | 3 - 4 | 406 | 67 | ||
M21476.1 | - | 1 - 2 | 105 | 67 | ||
X02918.1 | - | 3 - 4 | 528 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
