Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270939.1 | NP_001257868.1 | 1 - 2 | 57 | 73 | |
XM_006247867.2 | XP_006247929.1 | 1 - 2 | 86 | 73 | ||
XM_008768463.1 | XP_008766685.1 | 1 - 2 | 86 | 73 | ||
GenBank mRNA | CB792560.1 | - | 1 - 2 | 107 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
