Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270939.1 | NP_001257868.1 | 6 - 7 | 712 | 74 | |
NM_053969.3 | NP_446421.3 | 6 - 7 | 886 | 74 | ||
XM_006247865.1 | XP_006247927.1 | 6 - 7 | 881 | 74 | ||
XM_006247867.2 | XP_006247929.1 | 6 - 7 | 741 | 74 | ||
GenBank mRNA | BC061746.1 | - | 6 - 7 | 835 | 74 | |
X87885.1 | - | 5 - 6 | 667 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
