Assay Details
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053969.3 | NP_446421.3 | 1 - 2 | 235 | 99 | |
XM_006247865.1 | XP_006247927.1 | 1 - 2 | 230 | 99 | ||
XM_006247866.1 | XP_006247928.1 | 1 - 2 | 230 | 99 | ||
XM_008768462.1 | XP_008766684.1 | 1 - 2 | 230 | 99 | ||
GenBank mRNA | BC061746.1 | - | 1 - 2 | 184 | 99 | |
CB771566.1 | - | 1 - 2 | 235 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061746.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
