Assay Details
Gene Symbol: Ttc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005529.1 | NP_001005529.1 | 1 - 2 | 56 | 101 | |
XM_008767639.1 | XP_008765861.1 | 1 - 2 | 107 | 101 | ||
GenBank mRNA | BC082093.1 | - | 1 - 2 | 56 | 7134129 | 101 |
FQ213119.1 | - | 1 - 2 | 73 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC082093.1, NM_001005529.1, XM_008767639.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
