Assay Details
Gene Symbol: Eral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013229.1 | NP_001013247.1 | 5 - 6 | 603 | 65 | |
XM_008768087.1 | XP_008766309.1 | 5 - 6 | 599 | 65 | ||
GenBank mRNA | BC089885.1 | - | 5 - 6 | 603 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
